Learn More
SeaKem™ GTG™ Agarose, Lonza™
Standard gelling temperature. Lonza SeaKem™ GTG™ Agarose is specifically designed for preparative DNA electrophoresis and tested for presence of proteases, ligases and nucleases.
Brand:  Lonza 50075
Product Code. 11560727
  • 5260.00 EUR / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Description
  • High gel strength agarose that resolves DNA fragments greater than 1,000 bp
  • DNA recovered from agarose gels are reliably digested and ligated
  • Performance tested to ensure complete compatibility with routine molecular biology techniques
  • Certificate of performance containing end-use test information is provided
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 kg