Learn More
SeaKem™ LE Agarose, Lonza™
Click to view available options
Quantity:
25 g
100 g
125 g
500 g
Unit Size:
25g
500g
Each
Description
- For DNA analysis/blotting
- Molecular Biology Grade
- EEO 0.09–0.13
- Gelling temperature 36° ± 1.5°C
- DNase/RNase-free
Specifications
Specifications
| Chemical Name or Material | SeaKem LE Agarose |
| Quantity | 100 g |