Learn More
LABPLAS Secure-T Clear Open
Blender bag with open top designed to withstand the most demanding laboratory homogenizer tests.
Brand:  LABPLAS SCLO5512
 View more versions of this product
Product Code. 17950235
  • 303.00 EUR / Pack of 1000
Explore more special offers
Description
Description
  • Secure-T open sterile homogenizer bags are designed to withstand the most demanding laboratory tests.
  • Easy to use
  • Transparent to ensure full visibility of the sample
  • Available in multiple sizes
  • Thickness: 76.2 microns
Specifications
Specifications
Blender Bag
Clear
Sterile
Yes
3 mil.
5.5 in.
12 in.
Not Autoclavable
Single-use
76.2 μm
1000/Cs.
140 x 305 mm
140 mm
305 mm