Learn More
Brady™ Self-Laminating Rotating Vinyl Wrap Around Wire and Cable Labels for M6/M7 Printers
Identify your wires from any angle with Rotating Vinyl Wire Labels. Simply print your labels and then peel, fold and wrap – they’ll rotate freely for easy ID on even closed network connections.
Brand: Brady™ 174169
This item is not returnable.
View return policy
Description
- Rotates so you can identify your labels from any angle
- Simply wrap and apply--there's no adhesive residue or label parts to cling to your wires
- Spins freely without anchors so these labels won't damage your Cat 5 / 6 and fiber optic cables
- Made of B-427 vinyl
Specifications
Not Autoclavable | |
Permanent Acrylic | |
Clear, White | |
Die-cut | |
Single-side Printable | |
Labels | |
Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling | |
No | |
Rectangle | |
-70°C to +70°C | |
0.102 mm | |
2.75 in. | |
25.4 mm | |
25.4 mm | |
69.85 mm |
ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | |
None | |
Matte | |
Vinyl | |
Thermal Transfer | |
Non-sterile | |
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | |
50 Labels | |
-94°F to +158°F | |
0.004 in. | |
2.75 x 1 in. | |
1 in. | |
1 in. | |
1.1 in. | |
27.94 mm |