Learn More
Keystone Cleanroom Products Self-Sealing 1421B Tyvek™ Autoclave Bag with Steam Indicator, 24 to 62 in. Wide
The Self-Sealing 1421B Tyvek™ Autoclave Bag with Steam Indicator is designed for the steam sterilization of pharmaceutical equipment change parts. These bags are closed via the convenient and robust self-sealing closure system.
2515.00 EUR - 6325.00 EUR
Specifications
Product Type Autoclave Bag
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Color White
Disposable Single-use
View More Specs
Products 49
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
30020015
View Documents
Keystone Cleanroom Products
6STK2452SS
52 x 24 in.
6205.00 EUR
Pack of 250
30020018
View Documents
Keystone Cleanroom Products
6STK3434SS
34 x 34 in.
5410.00 EUR
Pack of 250
30020029
View Documents
Keystone Cleanroom Products
6STK2832SS
32 x 28 in.
4370.00 EUR
Pack of 250
30020037
View Documents
Keystone Cleanroom Products
6STK3412SS
12 x 34 in.
2955.00 EUR
Pack of 250
30020048
View Documents
Keystone Cleanroom Products
6STK5054SS
54 x 50 in.
2950.00 EUR
Pack of 100
30020075
View Documents
Keystone Cleanroom Products
6STK4416SS
16 x 44 in.
3775.00 EUR
Pack of 250
30020095
View Documents
Keystone Cleanroom Products
6STK2462SS
62 x 24 in.
6325.00 EUR
Pack of 250
30020096
View Documents
Keystone Cleanroom Products
6STK2844SS
44 x 28 in.
5530.00 EUR
Pack of 250
30020097
View Documents
Keystone Cleanroom Products
6STK4848SS
48 x 48 in.
3155.00 EUR
Pack of 125
30020101
View Documents
Keystone Cleanroom Products
6STK3016SS
16 x 30 in.
3670.00 EUR
Pack of 250
30020104
View Documents
Keystone Cleanroom Products
6STK3624SS
24 x 36 in.
4990.00 EUR
Pack of 250
30020108
View Documents
Keystone Cleanroom Products
6STK2634SS
34 x 26 in.
3995.00 EUR
Pack of 250
30020125
View Documents
Keystone Cleanroom Products
6STK4072SS
72 x 40 in.
4990.00 EUR
Pack of 125
30020132
View Documents
Keystone Cleanroom Products
6STK4040SS
40 x 40 in.
2945.00 EUR
Pack of 125
30020136
View Documents
Keystone Cleanroom Products
6STK3060SS
60 x 30 in.
2515.00 EUR
Pack of 100
30020140
View Documents
Keystone Cleanroom Products
6STK4864SS
64 x 48 in.
2950.00 EUR
Pack of 100
30020143
View Documents
Keystone Cleanroom Products
6STK2440SS
40 x 24 in.
4590.00 EUR
Pack of 250
30020145
View Documents
Keystone Cleanroom Products
6STK3672SS
72 x 36 in.
3565.00 EUR
Pack of 125
30020148
View Documents
Keystone Cleanroom Products
6STK3020SS
20 x 30 in.
3700.00 EUR
Pack of 250
30020170
View Documents
Keystone Cleanroom Products
6STK3628SS
28 x 36 in.
4990.00 EUR
Pack of 250
30020190
View Documents
Keystone Cleanroom Products
6STK4450SS
50 x 44 in.
3200.00 EUR
Pack of 125
30020197
View Documents
Keystone Cleanroom Products
6STK3030SS
30 x 30 in.
5010.00 EUR
Pack of 250
30020205
View Documents
Keystone Cleanroom Products
6STK2436SS
36 x 24 in.
4170.00 EUR
Pack of 250
30020208
View Documents
Keystone Cleanroom Products
6STK4060SS
60 x 40 in.
3565.00 EUR
Pack of 125
30020212
View Documents
Keystone Cleanroom Products
6STK3036SS
36 x 30 in.
5210.00 EUR
Pack of 250
30020226
View Documents
Keystone Cleanroom Products
6STK3664SS
64 x 36 in.
3200.00 EUR
Pack of 125
30020240
View Documents
Keystone Cleanroom Products
6STK4244SS
44 x 42 in.
3030.00 EUR
Pack of 125
30020248
View Documents
Keystone Cleanroom Products
6STK4850SS
50 x 48 in.
3200.00 EUR
Pack of 125
30020255
View Documents
Keystone Cleanroom Products
6STK3636SS
36 x 36 in.
5360.00 EUR
Pack of 250
30020270
View Documents
Keystone Cleanroom Products
6STK2828SS
28 x 28 in.
4345.00 EUR
Pack of 250
30020275
View Documents
Keystone Cleanroom Products
6STK3642SS
42 x 36 in.
2745.00 EUR
Pack of 125
30020278
View Documents
Keystone Cleanroom Products
6STK2646SS
46 x 26 in.
4090.00 EUR
Pack of 250
30020284
View Documents
Keystone Cleanroom Products
6STK3648SS
48 x 36 in.
2990.00 EUR
Pack of 125
30020292
View Documents
Keystone Cleanroom Products
6STK3646SS
46 x 36 in.
2855.00 EUR
Pack of 125
30020298
View Documents
Keystone Cleanroom Products
6STK3660SS
60 x 36 in.
3190.00 EUR
Pack of 125
30020299
View Documents
Keystone Cleanroom Products
6STK2424SS
24 x 24 in.
4545.00 EUR
Pack of 250
30020301
View Documents
Keystone Cleanroom Products
6STK2834SS
34 x 28 in.
4440.00 EUR
Pack of 250
30020302
View Documents
Keystone Cleanroom Products
6STK3028SS
28 x 30 in.
4640.00 EUR
Pack of 250
30020312
View Documents
Keystone Cleanroom Products
6STK2628SS
28 x 26 in.
3845.00 EUR
Pack of 250
30020339
View Documents
Keystone Cleanroom Products
6STK3838SS
38 x 38 in.
2855.00 EUR
Pack of 125
30020341
View Documents
Keystone Cleanroom Products
6STK3248SS
48 x 32 in.
3065.00 EUR
Pack of 125
30020342
View Documents
Keystone Cleanroom Products
6STK4048SS
48 x 40 in.
3305.00 EUR
Pack of 125
30020343
View Documents
Keystone Cleanroom Products
6STK4024SS
24 x 40 in.
2945.00 EUR
Pack of 125
30020350
View Documents
Keystone Cleanroom Products
6STK3456SS
56 x 34 in.
3200.00 EUR
Pack of 125
30020363
View Documents
Keystone Cleanroom Products
6STK4242SS
42 x 42 in.
2955.00 EUR
Pack of 125
30020364
View Documents
Keystone Cleanroom Products
6STK6250SS
50 x 62 in.
4090.00 EUR
Pack of 125
30020367
View Documents
Keystone Cleanroom Products
6STK2430SS
30 x 24 in.
3700.00 EUR
Pack of 250
30020379
View Documents
Keystone Cleanroom Products
6STK2448SS
48 x 24 in.
4990.00 EUR
Pack of 250
30020382
View Documents
Keystone Cleanroom Products
6STK3232SS
32 x 32 in.
5110.00 EUR
Pack of 250
Description
Description
  • Designed for steam sterilization
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Integrated steam indicator
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Eliminates need to cut material and the use of autoclave tape
  • Self-sealing closure is fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Resists tearing and punctures
  • Superior microbial barrier
  • Certificates of conformance sent with shipment
Specifications
Specifications
Autoclave Bag
Certificate of Conformance (COC)
Single-use
Pharmaceutical Equipment and Components
Non-sterile
Up to 250°F
5.3 mil
Autoclavable
White
Steam Sterilization
Tyvek
0.13 mm
Up to 125°C