Learn More
WTW™ SenTix™ 940-3 Digital IDS pH/ORP Electrodes
Click to view available options
pH Range:
0 to 14
Unit Size:
Each
Description
- Low-maintenance IDS pH gel electrode, 3 m fixed cable
- Digital measurement of pH with built-in electrode quality monitoring - usable in all areas of lab and field measurement
- Safely measured values due to signal processing in the sensor
- Easy electrode replacement thanks to calibration data stored inside sensor
- Sensor quality monitoring by QSC function that allows monitoring of the current sensor status with all IDS pH electrodes
- IDS pH electrodes for field use, and special applications in the lab.
- The QSC function allows monitoring of the current sensor status with all IDS pH electrodes
Specifications
Specifications
| Connector Types | Fixed Cable |
| Junction | Fiber |
| Material | Plastic Shaft, Fiber Diaphragm |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 80°C |
| Type | IDS pH/ORP Electrode |
| Includes | 3 m cable |
| Model | SenTix™ 940-3 |
| pH Relative Accuracy | ±0.004, ±1 |
| Probe | NTC |
| Show More |