missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Sephadex™ G-100, MP Biomedicals™

Product Code. 11475222 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11475222

Brand: MP Biomedicals™ 0219525580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in protein chromatography, affinity chromatography and gel filtration chromatography. Sephadex™ G-100, MP Biomedicals is a beaded gel filtration medium prepared by cross-linking dextran with epichlorohydrin under alkaline conditions.

This gel filtration medium is used in protein chromatography, affinity chromatography and gel filtration chromatography. Its main application is group separations of low and high molecular weight molecules. Desalting is the most common use.

  • Desalting with Sephadex is superior to dialysis because of the considerable time savings, the low dilution factor (which can be as low as 1.4 : 1), and the high activity recoveries even with very small amounts of sample
  • Related applications include buffer exchange and the removal of small molecules during the preparation of large biomolecules, such as ampholytes, detergents, radioactive or fluorescent labels, and phenol (during DNA purification).
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 g
Flow Rate >40ncm/hr.
Particle Size 40 to 120 μm
Product Type Sephadex G-100 Media
Format Free-flowing White Powder
Content And Storage Room Temperature (15–30°C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.