Learn More
Applied Biosystems™ Sequenase Version 2.0 DNA Polymerase
Sequenase™ Version 2.0 DNA Polymerase is a genetically engineered form of T7 DNA polymerase. Unlike the wild-type enzyme it has virtually no 3'→5' exonuclease activity.
230.00 EUR - 844.00 EUR
Specifications
Concentration | 13 units/μL |
---|---|
For Use With (Application) | Sequencing |
Polymerase | Genetically-Engineered form of T7 DNA Polymerase |
Product Type | Sequenase Version 2.0 DNA Polymerase |
Purity | 0.95 |
Product Code | Brand | Description | Purity | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Purity | Quantity | Price | Quantity & Availability | |||
15809896
|
Applied Biosystems™
70775Y200UN |
Sequenase Version 2.0 DNA Polymerase, 200 Units, 13 units/μL Concentration, Genetically-Engineered form of T7 DNA Polymerase, 7.5 pH, 37°C Optimum Temperature | 0.95 | 200 U |
230.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15819896
|
Applied Biosystems™
70775Z1000UN |
Sequenase Version 2.0 DNA Polymerase, 1000 Units, 13 units/μL Concentration, Genetically-Engineered form of T7 DNA Polymerase, 7.5 pH, 37°C Optimum Temperature | 0.95 | 1000 U |
844.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Specifications
13 units/μL | |
Genetically-Engineered form of T7 DNA Polymerase | |
0.95 |
Sequencing | |
Sequenase Version 2.0 DNA Polymerase |
For research use only