Learn More
TPP Techno Plastic Products™ Serological Pipets
Manufactured with the highest grade polystyrene for robustness and extreme break resistance. TPP Techno Plastic Products™ Serological Pipets offers large capacity volumes in a compact, ergonomic design.
Brand:  TPP Techno Plastic Products™ 94525
 View more versions of this product
Product Code. 11361784
  • 84.00 EUR / Pack of 60
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Description
Manufactured with the highest grade polystyrene for robustness and extreme break resistance. TPP Techno Plastic Products™ Serological Pipets offers large capacity volumes in a compact, ergonomic design.
  • Accurate volume; highly visible, bidirectional (ascending, descending); over-volume graduations
  • Smooth interiors, prevent cell hang-up or clumping inside the pipette
  • Excellent optical transparency
  • Color coded cotton plugs and dispenser boxes for easy identification
  • Convenient, stackable TPP dispenser box
  • Single wrapped in lint-free paper/plastic blister
  • Slender and short form enables ergonomic working conditions in the sterile hood
  • Small outside diameter of the tube at the tip results in an excellent accessibility of the large pipettes even in small tissue culture flasks
  • Highly drop free pipet tip design
  • Optimized form of the mouthpiece preserves the rubber of the pipet aid
  • Models available with reservoir; reservoir augments the total volume capacity without adding proportionate length to the pipet
Specifications
Specifications
Polystyrene
2/10
25 mL
Yes
Red