missing translation for 'onlineSavingsMsg'
Learn More

SERP1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA521385

Product Code. 13238938

  • 514.00 EUR / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Recommended positive controls: A431. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Pig (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene interacts with target proteins during their translocation into the lumen of the endoplasmic reticulum. Protects unfolded target proteins against degradation during ER stress. May facilitate glycosylation of target proteins after termination of ER stress. May modulate the use of N-glycosylation sites on target proteins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SERP1
Polyclonal
Unconjugated
SERP1
D3Ucla1; RAMP4; ribosome associated membrane protein 4; ribosome-attached membrane protein 4; Serp1; Stress-associated endoplasmic reticulum protein 1
Rabbit
Antigen affinity chromatography
RUO
27230, 28146
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Western Blot
1 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
Q9Y6X1, Q9Z1W5
SERP1
Synthetic peptide corresponding to a region within amino acids 1 and 51 of SERP1 (Uniprot ID#Q9Y6X1)
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers