Learn More
Trajan™ Replacement Needles for Luer-Lock Syringes
Product Code. p-4911131
Click to view available options
Needle Gauge:
14 G
16 G
18 G
19 G
23 G
Length (English) Needle:
2 in.
2.7 in.
4.52 in.
Unit Size:
Pack of 5
Product Code. 11989427
Brand: Trajan™ 039807
Bevel tip style. Trajan Scientific and Medical™ Replacement Needles for Luer-Lock Syringes are supplied in packs of 5.
Designed for optimum septum penetration and prevention of septum coring.
  • Standard general purpose needle
  • Made of stainless steel
  • Preferred for manual injection where piercing the septum in exactly the same place is difficult
Specifications
Material Stainless Steel
Needle Gauge 23 G
Diameter (English) Inner 0.012 in.
Diameter (English) Outer 0.024 in.
Diameter (Metric) Inner 0.32 mm
Diameter (Metric) Outer 0.63 mm
For Use With (Application) Gas chromatography
Length (English) Needle 2.7 in.
Product Type Replacement Needle
Product Line Trajan
Length (Metric) Needle 68.58 mm
Needle Point Style Bevel
Connection Type Luer Lock