Learn More
SGE™ Replacement Needles for 0.5 - 1μL Syringes
Product Code. 10472561
Click to view available options
Needle Gauge:
23 G
26 G
32 G
Needle Point Style:
Bevel
Cone
Sheathed/LC
Length (English) Needle:
2 in.
2.7 in.
4 in.
For Use With (Equipment):
8 mm O.D. Barrel 0.5μL Microvolume Syringes
8 mm O.D. Barrel 1μL Microvolume Syringes
Microvolume Syringes (1μL)
On-column Syringe
Syringe Needles (0.5μL), Syringes
Unit Size:
Each
Product Code. 10472561
Brand: SGE™ 034055
For use with 0.5 to 1μL syringes
For removable-needle syringes or additional replacements, contact your Fisher Scientific Customer Service Representative.
Specifications
Material Stainless Steel
Needle Gauge 23 G
Diameter (English) Inner 0.006 in.
Diameter (English) Outer 0.02 in.
Diameter (Metric) Inner 0.15 mm
Diameter (Metric) Outer 0.5 mm
Length (English) Needle 2 in.
Product Type Replacement Needle
Length (Metric) Needle 51 mm
Needle Point Style Cone
For Use With (Equipment) 8 mm O.D. Barrel 1μL Microvolume Syringes