Learn More
SPEX CertiPrep™ Mixer/Mill™ High-Energy Ball Mill
Shop All SPEX Certiprep Products
Click to view available options
Voltage:
230 V
Unit Size:
Each
Specifications
Specifications
| Dimensions (L x W x H) | 53 x 56 x 36 cm |
| Electrical Requirements | 230 V, 50 Hz |
| Horsepower | 1425 RPM @ 50Hz |
| Plug Type | EU plug |
| Speed | 875 cycles/min. |
| Voltage | 230 V |