Learn More
Epredia™ Cryochrome™ Embedding Resin
Product Code. 12726087 Shop All Epredia Products
Click to view available options
Product Type:
Cryochrome Blue
Cryochrome Green
Cryochrome Pink
Cryochrome Sampler Colors
Cryochrome Yellow
Unit Size:
120mL
Each
Pack of 4
Product Code. 12726087
Brand: Epredia™ 9990426
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Support tissue for frozen sectioning with Epredia™ Cryochrome™ Embedding Resin.
  • Color codes frozen sections for easy identification and tracking
  • Four colors available: pink, blue, yellow, green
  • Formulated with physical characteristics identical to Cryomatrix™ embedding resin
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cryochrome Sampler Colors
Quantity 120mL