Learn More
Epredia™ Shandon™ Embedding Cassettes II in Tube Packs, aqua
Hold tissue specimens during processing and embedding with Epredia™ Shandon™ Embedding Cassettes II in Tube Packs.
Brand:  Epredia™ B1000729GY
 View more versions of this product
Product Code. 12973755
Description
Description
  • Acetal polymer cassettes with writing face at 45° angle
  • Slotted pore style
  • Locking lid attached by two hinges
Ordering Information:
  • Tube packs are supplied in 10 tubes of 75 cassettes each
Specifications
Specifications
Slotted Pore
Gray
Biopsy Cassette
45°
Acetal Polymer