Learn More
Epredia™ Shandon™ Gram Stain
Differentiate between Gram-positive and Gram-negative bacteria with Epredia™ Shandon™ Gram Stain.
Brand: Epredia™ 9990726
Includes: Gram's iodine, 4L
Description
- Based upon the popular Hucker modification
- Demonstrates the presence of microorganisms such as bacteria
Specifications
Gram Iodine | |
Crystal violet iodine | |
4L |