Learn More
Epredia™ Shandon™ Gram Stain
Differentiate between Gram-positive and Gram-negative bacteria with Epredia™ Shandon™ Gram Stain.
Brand:  Epredia™ 9990726
 View more versions of this product
Product Code. 12599956
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Includes: Gram's iodine, 4L
Description
Description
  • Based upon the popular Hucker modification
  • Demonstrates the presence of microorganisms such as bacteria
Specifications
Specifications
Gram Iodine
Crystal violet iodine
4L