Learn More
Epredia™ Shandon™ Tissue-Processing Capsule, Separators, S-shaped, Capsule
Partition up to four sections of the Epredia™ Shandon™ Tissue Processing Capsule, constructed of stainless steel.
Description
Description
  • Five sturdy lugs grasp the base securely
  • Partition separators sold separately
Ordering Alert:
  • Dimensions (O.D. x D): 1.46 x 0.24 in. (37 x 6mm)