missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Mouse IgD Fc Secondary Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP2-69318
This item is not returnable.
View return policy
Description
IgD Fc Polyclonal antibody specifically detects IgD Fc in Mouse samples. It is validated for ImmunoprecipitationSpecifications
IgD Fc | |
Polyclonal | |
Unconjugated | |
Stable whole antiserum | |
Pools of purified homogenous IgD isolated from BALB/C and C57Bl mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
1 mL | |
Secondary | |
Mouse | |
Lyophilized |
Immunoprecipitation | |
LYOPH | |
Immunoprecipitation | |
Sheep | |
Whole Antiserum | |
RUO | |
Reconstitute by adding 1 mL sterile distilled water, spin down to remove insoluble particles. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction