missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Mouse IgG2a Secondary Antibody, Biotin, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP2-69567
This item is not returnable.
View return policy
Description
IgG2a Polyclonal antibody specifically detects IgG2a in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, Dot BlotSpecifications
IgG2a | |
Polyclonal | |
Biotin | |
Lyophilized from PBS (pH 7.2) | |
Highly purified normal homogenous IgG2a(1b) isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
1 mL | |
Secondary | |
Mouse | |
Lyophilized |
Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot | |
LYOPH | |
Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000 | |
Sheep | |
Ion exchange chromatography | |
RUO | |
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction