Learn More
Sheep anti-Mouse IgG2a Secondary Antibody, Biotin, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69570
Product Code. 18687376
  • 837.00 EUR / 1mL
Explore more special offers
Description
Description
IgG2a Polyclonal antibody specifically detects IgG2a in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, Dot Blot
Specifications
Specifications
IgG2a
Polyclonal
Biotin
Lyophilized from PBS (pH 7.2)
Highly purified normal homogenous IgG2a(1b) isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
1 mL
Secondary
Mouse
Lyophilized
Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot
LYOPH
Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000
Sheep
Ion exchange chromatography
RUO
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Store at 4°C in the dark.