Learn More
Emma Safety Footwear Shoe Laces
Available in White or black/gray colors.
4.20 EUR - 5.00 EUR
Product Code | Brand | Color | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Length (Metric) | Price | Quantity & Availability | ||||
10635015
|
Emma Safety Footwear
MM00006500 |
Black, Gray | 120 cm |
4.20 EUR
1 pair |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||||
10092154
|
Emma Safety Footwear
MM00006100 |
White | 95 cm |
5.00 EUR
1 pair |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
10566944
|
Emma Safety Footwear
MM00006400 |
Black, Gray | 95 cm |
4.75 EUR
1 pair |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||||