Learn More
Dräger™ Short-Term Detector Tubes - Perchloroethylene
Brand:  Dräger™ 8101551
 View more versions of this product
Product Code. 10570773
  • 615.00 EUR / Pack of 10
Explore more special offers
Description
Description
  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.
Specifications
Specifications
Perchloroethylene 0.1/a
0.5 to 4ppm
10
59°F to 86°F
30mg/L
3/9
3 to 9 min.
15°C to 30°C
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Perchloroethylene 0.1/a
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.