Learn More
Epredia™ Signature Series™ Hematoxylin 2
Product Code. p-4530224 Shop All Epredia Products
Click to view available options
Quantity:
1 pt.
Unit Size:
Pack of 4
Product Code. 10066478
Brand: Epredia™ 7231
Yield visually similar results to Gills 2 and 3 with this uniquely formulated product.
  • No filtration required
  • Stains up to 2500 slides per pint
  • Part of the Signature Series Staining System
  • Crisp, well-defined nuclear detail chromatin delineation
Available in 5L packaging
  • Reduces plastic by 90%
  • Increases stain stability
  • Reduces messy stain spills
  • Saves 35% storage space versus traditional packaging
TRUSTED_SUSTAINABILITY
Specifications
Type Hematoxylin 2
Quantity 1 pt.
compliance-icons
Product Identifier
  • X4 STAIN HEMATOXYLIN 2 4X1PT . .
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Ethylene glycol, Acetic acid