Learn More
DWK Life Sciences Silicone Sealing Ring, VMQ
The silicone seal is used for sealing and fixing threaded pipes with threaded caps.
Brand: DWK Life Sciences 292361407
103.55 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
The silicone sealing ring matches the screw connection piece and has good sealing properties. Due to the PTFE layer on one side of the sealing ring, there is a certain inertness of the material. The included hole allows the insertion of glass rods, olives, hoses, etc.
- With bonded PTFE face
Specifications
13.0 to 15.0 mm | |
GL 32 | |
Fits Tubing ID: 13.0 to 15.0 mm | |
Silicone (VMQ) |
Sealing; Connecting glass tubes; Suitable for PBT screw cap with aperture | |
10/Pk. | |
29 mm |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu