Learn More
Hirschmann™ SILICOSEN™ Pink Silicone Stoppers
Click to view available options
Diameter (Metric) Inner:
6-9 mm
9-11 mm
11-14 mm
13-16 mm
15-18 mm
18-21 mm
20-23 mm
22-30 mm
30-40 mm
40-50 mm
Length (Metric):
10 mm
12 mm
13 mm
15 mm
20 mm
30 mm
35 mm
40 mm
Unit Size:
Pack of 10
Description
- Uniform pore structure
- Good air permeability
- Chemically and temperature resistant
- Liquid repellent and reusable
Specifications
Specifications
| Diameter (Metric) Inner | 9-11 mm |
| For Use With (Application) | For The Preparation And Sterilization Of Culture Media |
| Length (Metric) | 13 mm |
| Quantity | 10/Pk. |
| Color | Pink |
| Material | Silicone |
| Product Type | Tapered Stopper |