Learn More
VistaLab™ Technologies Fixed Volume Ovation BioNatural Pipettes
Click to view available options
Volume (Metric):
5 μL
200 μL
300 μL
Unit Size:
Each
Description
- Aspirating and Dispensing: A very low-force,form-fitting plunger allows the user to manuallycontrol the speed of aspirating and dispensing with minimal contact or thumb effort.
- Tip Ejection: A low-force energy release button activates the tip ejector. A gentle push ensures that the forces needed to discard a tip are well below threshold levels recommended by ergonomists
- Tip Acquisition: The user inserts the nozzle into a tip and lightly presses down until it clicks. No excessive force or jamming is required to ensure a good seal
- Liquid Blocking Disposable Nozzle: Not every pipetting task requires a filtered tip, but every pipette should be protected from contamination no matter what the task. Ovation comes with a filtered nozzle that does not allow liquid into the body of the device
Specifications
Specifications
| Accuracy | ±1.9% |
| Color | Purple |
| Material | Polycarbonate ABS |
| Model | F1 |
| Number of Channels | 1 |
| Type | Fixed Volume Ovation Bionatural Pipette |
| Volume (Metric) | 300 μL |
| Precision | 0.4% |
| Quantity | 1 ea. |
| Warranty | 2 Year Limited Warranty |
| Show More |