Learn More
Six component organic solution, 1000mg/mL each, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical F112181
This item is not returnable.
View return policy
Description
Six component organic solution - Benzo(a)pyrene, Benzo(b)fluoranthene, Benzo(g,h,i)perylene, Benzo(k)fluoranthene, Fluoranthene, Indeno(1,2,3-c,d)pyrene 1,000mg/mL of eachSpecifications
Six Components PAH Standard Solution | |
Ampoule | |
Organic Multi-component Standard Solution | |
Acetonitrile |
Fluoranthene 1mg/mL, Benzo(b)fluoranthene 1mg/mL, Benzo(k)fluoranthene 1mg/mL, benzo(a)pyrene 1mg/mL, Benzo(g,h,i)perylene 1mg/mL, Indeno(1,2,3-c,d)pyrene 1mg/mL, Acetonitrile 1 part | |
1 mL | |
Fluoranthene, Benzo(b)fluoranthene, Benzo(k)fluoranthene, benzo(a)pyrene, Benzo(g,h,i)perylene, Indeno(1,2,3-c,d)pyrene, Acetonitrile |