missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A3 Rabbit anti-Human, Mouse, Clone: 1K0A4, Novus Biologicals™
Rabbit Monoclonal Antibody
191.14 EUR - 483.79 EUR
Specifications
Antigen | SLC22A3 |
---|---|
Clone | 1K0A4 |
Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18331356
|
Bio-Techne
NBP3-15897-20UL |
20 μg |
191.14 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18350413
|
Bio-Techne
NBP3-15897-100UL |
100 μg |
483.79 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC22A3 Monoclonal antibody specifically detects SLC22A3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SLC22A3 | |
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6581 | |
IgG | |
Affinity purified |
1K0A4 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
EMTEMTH, Extraneuronal monoamine transporter, OCT3EMT organic cation transporter 3, Organic cation transporter 3, solute carrier family 22 (extraneuronal monoamine transporter), member 3, solute carrier family 22 member 3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 301-450 of human SLC22A3 (O75751). GDKALQILRRIAKCNGKYLSSNYSEITVTDEEVSNPSFLDLVRTPQMRKCTLILMFAWFTSAVVYQGLVMRLGIIgGNLYIDFFISGVVELPGALLILLTIERLGRRLPFAASNIVAGVACLVTAFLPEGIAWLRTTVATLGRLGITMAF | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |