Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G
Product Code. 15860182 Shop All Thermo Scientific Products
Click to view available options
Description:
Magnetic, Soluble
Magnetic, Standalone
Magnetic, with SOLAμ Collection Plate, Soluble
Non-Magnetic, Soluble
Non-Magnetic, with SOLAμ Collection Plate, Soluble
Unit Size:
Each
Product Code. 15860182
Brand: Thermo Scientific™ 60115104
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific SMART Digest Bulk Fractionation Kits: Protein G.
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G. These kits remove the challenges often associated with protein cleanup by providing a simple and easy capture workflow.
SMART Digest Bulk Fractionation Kits: Protein G contain beads that target purification of immunoglobulins due to the specific and strong affinity for binding to the Fab and Fc portions of the molecule. Choose from non-magnetic beads, or magnetic beads which enable automation. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once complete, perform digestion using SMART Digest enzymes if required.
TRUSTED_SUSTAINABILITY
Specifications
Description Magnetic, Soluble
Capture Protein G
Cleavage C-terminal side of lysine and arginine amino acids
Enzyme Trypsin
Format Bulk Magnetic
For Use With (Application) Immunocapture and Protein Digest Sample Preparation
Product Line SMART Digest
Type SMART Digest Bulk Fractionation Kit
Includes Bulk Magnetic SMART Digest Soluble Resin