Learn More
Thermo Scientific™ Smart-Vue™ Sensors
Product Code. 11844703 Shop All Thermo Scientific Products
Click to view available options
Description:
Digital Sensor, 27cm cable length
Digital Sensor, 3.5m cable length
Digital Sensor; 1m cable length
Digital Temperature/RH Sensor
PT100 Temperature Sensor, 1.4m cable length
Temperature/CO2 Sensor, 1m cable length
Calibration:
-30, -15, 0, +20, +40°C
23°C, 50% RH
30, 60, 80, 100, 105°C
37°C, 5% CO2
Not Calibrated
Unit Size:
Each
Product Code. 11844703
Brand: Thermo Scientific™ SV400100LSB
Protect the quality of your samples, facilitate regulatory compliance and continuously monitor and report conditions using Thermo Scientific™ Smart-Vue™ Sensors.
Protect the quality of your samples, facilitate regulatory compliance and continuously monitor and report conditions using Thermo Scientific™ Smart-Vue™ Sensors. Monitor temperatures ranging from -200° to +350°C, CO2 concentration, relative humidity (RH), differential pressure (clean rooms), 4-20 milliamps and dry contact state.
  • Automatic and continuous data logging
  • Temperature, temperature/CO2, temperature/rH or temperature/CO2/rH combo sensors
Compatible with:
Refrigerators, Freezers, LN2 Storage, CO2 Incubators, Clean Rooms, Storage Areas, Ovens
Recommended for:
Ideal for use in highly controlled environments, including pharmaceutical production, biotechnology, IVF clinics, clinical studies and diagnostic laboratories.
Ordering Information:
Availability of Smart-Vue products vary by country. Please ask your Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.
Specifications
Type Sensor
Calibration -30, -15, 0, +20, +40°C
Description Digital Sensor; 1m cable length
Certifications/Compliance Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Application) Cold Storage
For Use With (Equipment) Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Temperature Range (Metric) -40°C to +80°C