missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Smoothened Agonist Dihydrochloride Dihydrate
A highly potent activator of Smoothened
Chemical Identifiers
CAS | 364590-63-6 |
---|---|
Molecular Formula | C28H28ClN3OS.2HCl.2H2O, C28H32Cl3N3O2S |
Synonym | SAG1.3 HCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15426549
|
Thermo Scientific Chemicals
J64726.M^ |
1 mg |
N/A
|
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15416549
|
Thermo Scientific Chemicals
|
5 mg |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
364590-63-6 | |
C28H28ClN3OS.2HCl.2H2O, C28H32Cl3N3O2S | |
SAG1.3 HCl |
Specifications
Pale yellow | |
A highly potent activator of Smoothened | |
C28H28ClN3OS.2HCl.2H2O, C28H32Cl3N3O2S | |
SAG1.3 HCl | |
Dihydrochloride Dihydrate | |
IYXUQUYRWPGIQL-UHFFFAOYSA-N | |
3-chloro-N-[4-(methylamino)cyclohexyl]-N-{[3-(pyridin-4-yl)phenyl]methyl}-1-benzothiophene-2-carboxamide hydrate dihydrochloride |
2°C to 8°C | |
364590-63-6 | |
599.00 | |
Light Sensitive and Hygroscopic | |
Solid | |
O.Cl.Cl.CNC1CCC(CC1)N(CC1=CC(=CC=C1)C1=CC=NC=C1)C(=O)C1=C(Cl)C2=CC=CC=C2S1 | |
580.99 |
Safety and Handling
Recommended Storage : Keep cold
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title