Learn More
Sampling Systems™ Snapper Sediment Sampler Telescopic Rod
The Snapper sediment sampler is ideal for taking samples of earth, silt and small stones in a variety of different environmental settings such as rivers, lakes and ponds.
Brand: Sampling Systems™ 5061T-4500
Additional Details : Weight : 1.00000kg
Description
- Can be used in conjunction with telescopic rods
- Spring loaded mechanism closes when it hits firm ground or is given a sharp jolt on the handle
Specifications
Telescopic Rod | |
Not Autoclavable | |
No |
Aluminum | |
Reusable | |
Non-sterile |