missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS3 Mouse anti-Human, Unconjugated, Clone: 1G2B12, Proteintech
Mouse Monoclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | SOCS3 |
---|---|
Clone | 1G2B12 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16829005
|
Proteintech
66797-1-IG-20UL |
20 μL |
140.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16819005
|
Proteintech
66797-1-IG-150UL |
150 μL |
399.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SOCS3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SOCS3. This antibody recognizes human, mouse antigen. The SOCS3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
SOCS3 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
9021 | |
Primary |
1G2B12 | |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Mouse | |
IgG1 |