missing translation for 'onlineSavingsMsg'
Learn More

Soda Lime Glass Centrifuge Tubes with Rim

Use these tubes for a variety of applications. Soda Lime Glass Centrifuge Tubes provide excellent sample retention during centrifugation, and fill rapidly without affecting specimen integrity.

Brand:  Samco Y138/71

 View more versions of this product

Product Code. 12853472

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Soda glass
  • Capacity 15mL
  • Max. RCF x g: 1300
Specifications

Specifications

15 mL
10
Yes
Centrifuge Tube
Soda Lime Glass
Cylindrical
Yes
1300 x g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers