Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Culture Tube
Product Code. 11892393
Click to view available options
Length (Metric):
40 mm
50 mm
55 mm
70 mm
120 mm
125 mm
150 mm
Diameter (Metric):
8 mm
9.5 mm
10.5 mm
11 mm
12 mm
16 mm
Unit Size:
Pack of 200
Pack of 250
Pack of 500
Product Code. 11892393
Brand: DWK Life Sciences A405511000811
Use the Soda Lime Glass Culture Tube to view cell cultures in the laboratory. This disposable soda lime glass rim top tube is plain (unlabeled) and features a sturdy, uniform bottom.
  • Rimmed top
  • Unlableled
  • Clear soda lime glass
  • Uniform bottom
  • Disposable
TRUSTED_SUSTAINABILITY
Specifications
Design Type Round bottom
Length (Metric) 55 mm
Packaging 500Pack
Diameter (Metric) 11 mm
Product Type Test Tube