Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Culture Test Tubes
Click to view available options
Length (Metric):
50 mm
75 mm
100 mm
160 mm
180 mm
Diameter (Metric):
6.5 mm
10 mm
11.5 mm
12 mm
13 mm
16 mm
18 mm
Unit Size:
Pack of 100
Pack of 1000
Pack of 250
Description
- Soda lime glass
- Round bottom
- X250
Specifications
Specifications
| Design Type | Round-bottom |
| Description | Test Tube |
| Length (Metric) | 100 mm |
| Material | Soda Lime Glass |
| Quantity | 250 Pack |
| Autoclavable | Autoclavable |
| Diameter (Metric) | 12 mm |