Learn More
Sodium acetate, 3M aq. soln., pH 4.5, autoclaved, Thermo Scientific Chemicals
Product Code. 15400187
Click to view available options
Quantity:
125 mL
250 mL
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O
Specifications
Linear Formula NaOOCCH3
Concentration 3M aq. soln.
IUPAC Name sodium;acetate
Formula Weight 82.03
Odor Odorless
pH 4.5
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Sodium acetate, autoclaved