missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, 99.5% min., (USP), MP Biomedicals

Product Code. 11496272
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
1 kg
10 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Quantity 1 kg
Assay Percent Range 99.5% min.
Formula Weight 58.44
Grade USP
Chemical Name or Material Sodium Chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'