missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, ULTREX™ Ultrapure Reagent, J.T. Baker™

Product Code. 15518324 Shop All J.T.Baker Products
Click to view available options
Quantity:
100 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15518324

Brand: Avantor J.T.Baker 492405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7647-14-5
Quantity 100 g
Molecular Formula ClNa
MDL Number MFCD00003477
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Formula Weight 58.44
Grade ULTREX™ Ultrapure Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.16kg
Chemical Name or Material Sodium Chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.