missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium dodecyl sulphate (SDS), BAKER, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
:
1 kg
Specifications
Specifications
| Melting Point | 204°C to 207°C |
| CAS | 151-21-3 |
| Quantity | 1 kg |
| Assay Percent Range | min. 95% |
| Molecular Formula | C12H25NaO4S |
| MDL Number | MFCD00036175 |
| Flash Point | >100°C |
| Synonym | Lauryl sulphate sodium salt , Dodecyl sulphuric acid sodium salt , Sodium lauryl sulphate , Dodecyl sulphate sodium salt |
| InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
| SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362-Take off contaminated clothing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction