Learn More
Sodium formate, ACS, 99.0% min, Thermo Scientific Chemicals
71.30 EUR - 253.00 EUR
Chemical Identifiers
CAS | 141-53-7 |
---|---|
Molecular Formula | CHNaO2 |
Molecular Weight (g/mol) | 68.007 |
MDL Number | MFCD00013101 |
InChI Key | HLBBKKJFGFRGMU-UHFFFAOYSA-M |
Synonym | sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744 |
PubChem CID | 2723810 |
ChEBI | CHEBI:62965 |
IUPAC Name | sodium;formate |
SMILES | C(=O)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387888
|
Thermo Scientific Alfa Aesar
036424.36 |
500 g |
71.30 EUR
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377888
|
Thermo Scientific Alfa Aesar
036424.A3 |
2 kg |
253.00 EUR
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reducing agent, buffering agent, complexing agent, Noble metal precipitantSodium formate is used in analytical chemistry to precipitate noble metals. It is used for tanning leather, for dyeing and printing fabrics. It acts as an electroplating agent, an acidulant in the textile industry, a reducing agent, a mordant, a complexing agent. Further, it is used in photographic fixing baths. In addition, it is used as a buffering agent for strong mineral acids and food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141-53-7 | |
68.007 | |
HLBBKKJFGFRGMU-UHFFFAOYSA-M | |
2723810 | |
sodium;formate |
CHNaO2 | |
MFCD00013101 | |
sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744 | |
CHEBI:62965 | |
C(=O)[O-].[Na+] |
Specifications
141-53-7 | |
1.92 g/mL | |
CHNaO2 | |
3595134 | |
sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744 | |
HLBBKKJFGFRGMU-UHFFFAOYSA-M | |
sodium;formate | |
2723810 | |
68.01 | |
Crystalline |
259°C to 262°C | |
0.99 | |
MFCD00013101 | |
Hygroscopic | |
Soluble in water,glycerol. Slightly soluble in alcoholSoluble in water,alcohol,glycerol and formic acid. Insoluble in ether. | |
C(=O)[O-].[Na+] | |
68.007 | |
CHEBI:62965 | |
ACS Reagent | |
Sodium formate, For analysis ACS |
Safety and Handling
P210-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H320-H335-H500
missing translation for 'einecsNumber' : 205-488-0
missing translation for 'rtecsNumber' : LR0350000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only