missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide 30% w/w (40% w/v), Reagecon
Sodium Hydroxide 30% w/w (40% w/v)
Brand: Reagecon S30WW1
This item is not returnable.
View return policy
Description
- Sodium Hydroxide 30% w/w (40% w/v)
Specifications
30% w/w (40% w/v) | |
1 L | |
HDPE Bottle GL45 | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- SODIUM HYDROXIDE 30% w/w
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide