missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 5M/l Solution, Scharlab

Product Code. 11338514
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in analytical chemistry as a laboratory reagent and a titrant in volumetric analysis.

This volumetric solution was checked by means of potentiometric methods using Scharlau's potassium hydrogen phthalate volumetric standard. Scharlau's volumetric standards are directly traceable to the Standard Reference Materials from NIST (National Institute of Standards and Technology, USA).

  • Molecular Formula: NaOH
  • Molar Mass: 40.00g/mol
  • CAS: 1310-73-2
  • EIN ECS No.: 215-185-5
  • Factor: 0.000–1.001

Specifications

CAS 1310-73-2
Molecular Formula HNaO
Concentration or Composition Notes 1liter = 200.0g (5 mol) NaOH
Density ∼1.18g/cm3 
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Normality 5.0N
Traceability to NIST Traceable to NIST
Physical Form Solution
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide, solution 5 mol/l (5 N)
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'