missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide solution Strong EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12980534 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
This item is not returnable. View return policy

Product Code. 12980534

Brand: Fisher Chemical RP 200 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging Bottle
Analyte or Component Names Sodium Hydroxide
Purity Grade Notes EU Ph Ref: 1081404
Element Sodium Hydroxide
Quantity 1000 mL
Grade Eu. Ph.
compliance-icons
Product Identifier
  • Sodium Hydroxide Solution,Strong. European Pharmacopoeia Ref: 1081404 (RP 200)
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.