missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodate, puriss p.a., ≥99.5% (RT), Honeywell™ Fluka™

Product Code. 15676830 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 15676830

Brand: Honeywell-Fluka 7170225G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a.,≥99.5% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 7681-55-2
Quantity 25 g
Molecular Formula INaO3
Linear Formula NaIO3
MDL Number MFCD00003533
UN Number UN1479
Synonym sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat
InChI Key WTCBONOLBHEDIL-UHFFFAOYSA-M
SMILES [O-]I(=O)=O.[Na+]
IUPAC Name sodium;iodate
Molecular Weight (g/mol) 197.891
PubChem CID 23675764
ChEBI CHEBI:81708
Formula Weight 197.89g/mol
Percent Purity ≥99.5% (RT)
Packaging Glass bottle
Density 4.28g/mL at 25°C
Chemical Name or Material Sodium iodate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.