missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite Solution, Volumetric, 0.1M NaNO2 (0.1N), Honeywell™ Fluka™

Product Code. 15626750 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 15626750

Brand: Honeywell-Fluka 707761L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Volumetric, 0.1 M NaNO2 (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 7632-00-0
Quantity 1 L
Molecular Formula NNaO2
Linear Formula NaNO2
MDL Number MFCD00011118
UN Number NONH for all modes of transport
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
SMILES [Na+].[O-]N=O
IUPAC Name sodium nitrite
Molecular Weight (g/mol) 69.00
PubChem CID 23668193
ChEBI CHEBI:78870
Formula Weight 69g/mol
Packaging Glass bottle
Chemical Name or Material Sodium nitrite solution, Volumetric, 0.1M NaNO2 (0.1N)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.