missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic heptahydrate, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
4 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 4 x 2.5 kg |
Linear Formula | Na2HPO4 · 7H2O |
IUPAC Name | disodium phosphoric acid heptahydrate |
Formula Weight | 268.07 |
Percent Purity | 98.0 to 102.0% |
Density | 1.68g/mL at 25°C; Vapor Density: 4.9 (vs air) |
Chemical Name or Material | Sodium phosphate dibasic heptahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'