missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic heptahydrate, Honeywell Fluka™

Product Code. 15628140
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
4 x 2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS reagent, 98.0-102.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H17Na2O11P
Molecular Weight (g/mol) 270.08
MDL Number MFCD00149180
InChI Key PYLIXCKOHOHGKQ-UHFFFAOYSA-N
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
PubChem CID 6096963
SMILES O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O

Specifications

Quantity 6 x 500 g
Linear Formula Na2HPO4 · 7H2O
IUPAC Name disodium;hydrogen phosphate;heptahydrate
Formula Weight 268.07
Percent Purity 98.0 to 102.0%
Density 1.68g/mL at 25°C; Vapor Density: 4.9 (vs air)
Chemical Name or Material Sodium phosphate dibasic heptahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'