Learn More
Sodium succinate hexahydrate, Thermo Scientific Chemicals
45.50 EUR - 131.00 EUR
Chemical Identifiers
CAS | 6106-21-4 |
---|---|
Molecular Formula | C4H16Na2O10 |
Molecular Weight (g/mol) | 270.142 |
MDL Number | MFCD00149117 |
InChI Key | ZBTUYCUNQBRXOR-UHFFFAOYSA-L |
Synonym | disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate |
PubChem CID | 3083938 |
ChEBI | CHEBI:63686 |
IUPAC Name | disodium;butanedioate;hexahydrate |
SMILES | C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11357718
|
Thermo Scientific Alfa Aesar
033386.30 |
250 g |
45.50 EUR
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347718
|
Thermo Scientific Alfa Aesar
033386.A1 |
1 kg |
131.00 EUR
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium succinate hexahydrate is used as an ingredient of topical preparations for the treatment of cataract.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6106-21-4 | |
270.142 | |
ZBTUYCUNQBRXOR-UHFFFAOYSA-L | |
3083938 | |
disodium;butanedioate;hexahydrate |
C4H16Na2O10 | |
MFCD00149117 | |
disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate | |
CHEBI:63686 | |
C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+] |
Specifications
6106-21-4 | |
C4H16Na2O10 | |
MFCD00149117 | |
Hygroscopic | |
disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate | |
ZBTUYCUNQBRXOR-UHFFFAOYSA-L | |
disodium;butanedioate;hexahydrate | |
3083938 | |
270.14 (162.05 Anhydrous) | |
Sodium succinate hexahydrate |
120°C-6H2O | |
NaO2CCH2CH2CO2Na·6H2O | |
3921044 | |
14,8679 | |
It is soluble in water at a concentration of 270g/L at 20°C (68°F) . | |
C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+] | |
270.142 | |
CHEBI:63686 | |
Crystalline or Granular Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 205-778-7
missing translation for 'rtecsNumber' : WM7751000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only